Skip to Content
Merck
All Photos(1)

Key Documents

34238

Supelco

Bismerthiazol

PESTANAL®, analytical standard

Synonym(s):

N,N′-Methylene-bis(2-amino-5-sulfhydryl-1,3,4-thiadiazole)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N6S4
CAS Number:
Molecular Weight:
278.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

194-198 °C

suitability

complies for identity (by LC-MS)

application(s)

agriculture
environmental

format

neat

SMILES string

S=C1NN=C(NCNC2=NNC(=S)S2)S1

InChI

1S/C5H6N6S4/c12-4-10-8-2(14-4)6-1-7-3-9-11-5(13)15-3/h1H2,(H,6,8)(H,7,9)(H,10,12)(H,11,13)

InChI key

RSNWORHVUOZYLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-tao Wang et al.
Sichuan da xue xue bao. Yi xue ban = Journal of Sichuan University. Medical science edition, 35(6), 770-773 (2004-12-03)
To study the effect of pesticide N'N-methylene-bis on thyroid function in rats, and the time-effect relationship. N'N-methylene-bis dimethylsulfoxide solution was administered to rats by gavage at a dose of 50 mg/kg body weight for 1, 3, and 5 days respectively.
Yong-Jie Qiu et al.
Sichuan da xue xue bao. Yi xue ban = Journal of Sichuan University. Medical science edition, 41(1), 73-76 (2010-04-08)
To study the impact of N' N-methylene-bis on thyroglobulin produced by FRTL-5 cells, and to explore the potential of using FRTL-5 cells to screen environmental thyroid hormone disruptors in vitro. The FRTL-5 cells were treated with 0.1, 1.0 and 10.0
Weibo Cheng et al.
Sichuan da xue xue bao. Yi xue ban = Journal of Sichuan University. Medical science edition, 34(3), 472-475 (2003-08-13)
To study the biological effects of thyroid induced by N',N-methylene-bis (2-amino-5-sulfhydryl-1, 3, 4-thiadianole), discussing whether this agent is a thyroid hormone disruptor. Sixty Wistar rats were divided into 2 groups, including one experiment group and one control. The rats of
[Short-term cell transformation tests for N,N'-methylene-bis(2-amino-1,3,4-thiadiazole), N,N'-methylene-bis- (2-amino-5-sulfhydryl-1,3,4-thiazole) and relation with sister chromatid exchange and chromosome aberration].
Z D Fu
Zhonghua yu fang yi xue za zhi [Chinese journal of preventive medicine], 18(3), 146-150 (1984-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service