Skip to Content
Merck
All Photos(1)

Key Documents

34178

Supelco

Mesosulfuron-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21N5O9S2
CAS Number:
Molecular Weight:
503.51
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1ccc(CNS(C)(=O)=O)cc1S(=O)(=O)NC(=O)Nc2nc(OC)cc(OC)n2

InChI

1S/C17H21N5O9S2/c1-29-13-8-14(30-2)20-16(19-13)21-17(24)22-33(27,28)12-7-10(9-18-32(4,25)26)5-6-11(12)15(23)31-3/h5-8,18H,9H2,1-4H3,(H2,19,20,21,22,24)

InChI key

NIFKBBMCXCMCAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Mesosulfuron-methyl is a sulfonylurea herbicide, which is used in controlling ryegrass in wheat and broad-leaved weeds in other cereals.[1][2]

Application

Mesosulfuron-methyl may be used as a reference standard for the analysis of mesosulfuron-methyl herbicide in water samples using solid-phase extraction with liquid chromatography coupled to tandem mass spectrometry (SPE-LC-MS/MS).[3]
Mesosulfuron-methyl may be used as a reference standard for the determination of the analyte in environmental matrices by solid-phase extraction and solid-liquid extraction followed by liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).[4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of 12 sulfonylurea herbicides in drinking water after SPE by LC-DAD.
Gallitzendorfer R, et al.
Chromatographia, 73(7-8), 813-816 (2011)
Efficiency of mesosulfuron-methyl and clodinafop-propargyl dose for the control of Lolium perenne in wheat.
Vazan S, et al.
Crop Protection, 30(6), 592-597 (2011)
Yong In Kuk et al.
Pest management science, 63(4), 349-357 (2007-02-23)
Diclofop-resistant Lolium species (ryegrass) is a major weed problem in wheat production worldwide. This study was conducted to determine the resistance pattern of diclofop-resistant ryegrass accessions from the southern United States to mesosulfuron-methyl, a recently commercialized herbicide for ryegrass control
Christina T Echagarruga et al.
eLife, 9 (2020-10-06)
Cortical neural activity is coupled to local arterial diameter and blood flow. However, which neurons control the dynamics of cerebral arteries is not well understood. We dissected the cellular mechanisms controlling the basal diameter and evoked dilation in cortical arteries
Angélique Lazartigues et al.
Talanta, 85(3), 1500-1507 (2011-08-03)
Pesticides residues in aquatic ecosystems are an environmental concern which requires efficient analytical methods. In this study, we proposed a generic method for the quantification of 13 pesticides (azoxystrobin, clomazone, diflufenican, dimethachlor, carbendazim, iprodion, isoproturon, mesosulfuron-methyl, metazachlor, napropamid, quizalofop and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service