Skip to Content
Merck
All Photos(1)

Documents

33875

Supelco

Boscalid

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-N-(4′-chlorobiphenyl-2- yl)-nicotinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12Cl2N2O
CAS Number:
Molecular Weight:
343.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

form

solid

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(cc1)-c2ccccc2NC(=O)c3cccnc3Cl

InChI

1S/C18H12Cl2N2O/c19-13-9-7-12(8-10-13)14-4-1-2-6-16(14)22-18(23)15-5-3-11-21-17(15)20/h1-11H,(H,22,23)

InChI key

WYEMLYFITZORAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boscalid is a fungicide, which belongs to the carboxamide class of fungicides. Its mode of action involves binding to the enzyme succinate dehydrogenase in mitochondria and thus inhibits the fungal respiration.

Application

Boscalid may be used as a reference standard in the determination of boscalid in fruits and vegetable samples using ultra performance liquid chromatography coupled with tandem mass spectrometry (UPLC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Microbial Plant Pathogens: Detection and Management in Seeds and Propagules (2016)
Resistance to boscalid fungicide in Alternaria alternata isolates from pistachio in California.
Avenot FH and Michailides JT
Plant Disease, 91(10), 1345-1350 (2007)
Determination of succinate-dehydrogenase-inhibitor fungicide residues in fruits and vegetables by liquid chromatography?tandem mass spectrometry.
Abad-Fuentes A, et al.
Analytical and Bioanalytical Chemistry, 407(14), 4207-4211 (2015)
Reed M Johnson et al.
PloS one, 8(1), e54092-e54092 (2013-02-06)
Chemical analysis shows that honey bees (Apis mellifera) and hive products contain many pesticides derived from various sources. The most abundant pesticides are acaricides applied by beekeepers to control Varroa destructor. Beekeepers also apply antimicrobial drugs to control bacterial and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service