Skip to Content
Merck
All Photos(1)

Key Documents

32757

Supelco

Aflatoxin G2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Beilstein:
1692017
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

[H][C@@]12CCO[C@]1([H])Oc3cc(OC)c4C5=C(C(=O)OCC5)C(=O)Oc4c23

InChI

1S/C17H14O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WPCVRWVBBXIRMA-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Warning

Possibly carcinogenic.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Carc. 1B - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert A Everley et al.
Journal of analytical toxicology, 31(3), 150-156 (2007-06-21)
Automated immunoaffinity solid-phase extraction followed by liquid chromatography-tandem mass spectrometry and chemical analogue internal standardization is employed to detect and quantify the aflatoxins AFB(1), AFB(2), AFG(1), AFG(2), and the metabolites AFM(1) and AFP(1) in urine. The dynamic range of the
J Tam et al.
Food additives and contaminants, 23(7), 693-699 (2006-06-06)
Three hundred and forty-nine breakfast and infant cereal samples were collected at retail level across Canada from 2002 to 2005. They included rice-, soy-, barley-based and mixed-grain infant cereals, corn-, wheat-, rice-based and mixed-grain breakfast cereals, and were analysed for
Viresh Mohanlall et al.
Journal of food protection, 69(9), 2224-2229 (2006-09-26)
Phytoalexins (stress-induced compounds) from Citrus sinensis cultivar Valencia were screened for antifungal and antimycotoxic activity against a test organism (Cladosporium cladosporoides) and mycotoxin-producing fungi Fusarium verticillioides and Aspergillus parasiticus. The active compound, a member of the coumarin family of compounds
M Mably et al.
Food additives and contaminants, 22(12), 1252-1257 (2005-12-17)
Between March 1998 and March 2002, 304 samples of domestic (Canadian) and imported beers from 36 countries were picked up for the determination of aflatoxins B1, B2, G1 and G2. Twelve samples were positive with aflatoxins greater than the limit
Kathleen D'Ovidio et al.
Food additives and contaminants, 23(2), 174-180 (2006-02-02)
Ginseng roots can be infected by molds during growth, harvest and storage and result in contamination with mycotoxins. In this study, an analytical method for the determination of aflatoxins B(1), B(2), G(1) and G(2), a group of structurally similar mycotoxins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service