Skip to Content
Merck
All Photos(3)

Key Documents

294772

Sigma-Aldrich

Decahydronaphthalene, mixture of cis + trans

anhydrous, ≥99%

Synonym(s):

Decalin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18
CAS Number:
Molecular Weight:
138.25
Beilstein:
878165
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
189-191 °C (lit.)
vapor pressure:
42 mmHg ( 92 °C)
741 mmHg ( 188 °C)

grade

anhydrous

Quality Level

vapor density

4.76 (vs air)

vapor pressure

42 mmHg ( 92 °C)
741 mmHg ( 188 °C)

Assay

≥99%

form

liquid

autoignition temp.

482 °F

expl. lim.

0.7-4.9 %, 100 °F

impurities

<0.002% water
<0.005% water (100 mL pkg)

refractive index

n20/D 1.474 (lit.)

bp

189-191 °C (lit.)

mp

−125 °C (lit.)

density

0.896 g/mL at 25 °C (lit.)

SMILES string

C1CCC2CCCCC2C1

InChI

1S/C10H18/c1-2-6-10-8-4-3-7-9(10)5-1/h9-10H,1-8H2

InChI key

NNBZCPXTIHJBJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Decahydronaphthalene, mixture of cis + trans has been used as a liquid phase to study the reaction rate of hydrogenation of cyclohexene in the presence of a Pd–Al/biomorphic carbon catalyst.

Legal Information

Decalin is a trademark of Sigma-Aldrich Co. LLC

recommended

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1C

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of liquid phase cyclohexene hydrogenation on Pd?Al/biomorphic carbon catalysts.
Caza?a F, et al.
Catalysis Today, 249, 127-136 (2015)
Poly (diketopyrrolopyrrole-benzothiadiazole) with Ambipolarity Approaching 100% Equivalency.
Cho S, et al.
Advances in Functional Materials, 21(10), 1910-1916 (2011)
Manuel F Wagenhofer et al.
Science advances, 6(19), eaax5331-eaax5331 (2020-05-20)
Unsupported Ni-Mo sulfides have been hydrothermally synthesized and purified by HCl leaching to remove Ni sulfides. Unblocking of active sites by leaching significantly increases the catalytic activity for dibenzothiophene hydrodesulfurization. The site-specific rates of both direct (hydrogenolytic) and hydrogenative desulfurization
The Densities and Surface Tensions of cis-and trans-Decahydronaphthalene between-30 and 180?1.
Seyer WF and Davenport CH.
Journal of the American Chemical Society, 63(9), 2425-2427 (1941)
Conformational isomerization of cis-decahydronaphthalene over zeolite catalysts.
Lai WC and Song C.
Catalysis Today, 31(1), 171-181 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service