Skip to Content
Merck
All Photos(3)

Key Documents

195758

Sigma-Aldrich

Tetramethylammonium bromide

98%

Synonym(s):

TMAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(Br)
CAS Number:
Molecular Weight:
154.05
Beilstein:
3620955
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Quality Level

Assay

98%

form

solid

mp

>300 °C (lit.)

functional group

amine

SMILES string

[Br-].C[N+](C)(C)C

InChI

1S/C4H12N.BrH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1

InChI key

DDFYFBUWEBINLX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tetrabutylammonium bromide is a quaternary ammonium compound that is widely used as a phase transfer catalyst. Absolute viscosities of its aqueous solutions have been measured at various temperatures (20,25 and 30°C). Various physical properties (activity coefficient up to saturation, conductance, density and solubility in water) of TMABr have been evaluated.
Tetramethylammonium bromide (TMABr) is a quaternary ammonium salt that is widely used as a phase transfer catalyst. Absolute viscosities of its aqueous solutions have been measured at various temperatures (20,25 and 30°C). Various physical properties (activity coefficient up to saturation, conductance, density and solubility in water) of TMABr have been evaluated. Its impact on the first order rate constant values for the base hydrolysis of the following Fe(II) chelates has been investigated:




  • bis(naphthylidene alanate) (nali)
  • bis(naphthylidene phenylalanate) (nphali)
  • bis(naphthylidene aspartate) (nasi)
  • (naphthylidene histidinate) (nhi)
  • bis(naphthylidenearginate) (nari)

Application

Tetrabutylammonium bromide may be used in the molten state in the following processes:
  • Synthesis of (2S)-5-(3-phenyl-2-phthalimidylpropanoylamino)isophthalic acid.
  • Synthesis of alkyl-substituted pyrroles in the absence of catalyst and organic solvent.
  • Synthesis of dithioacetals from acetals by transthioacetalisation in a solvent free environment.
  • Synthesis of polyamides (PAs) by the polymerization of terephthalic acid and diisocyanates.
  • Catalyze the addition of thiols to conjugated alkenes.
  • Dehydrochlorination of poly(vinyl chloride).
Tetramethylammonium bromide may be used as a template in the synthesis of small colloidal zeolite Y nanocrystals.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlling size and yield of zeolite Y nanocrystals using tetramethylammonium bromide.
Holmberg BA, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 59(1), 13-28 (2003)
Viscosity of aqueous solutions. III. Tetramethylammonium bromide and the role of the tetraalkylammonium ions.
Nightingale Jr ER.
The Journal of Physical Chemistry, 66(5), 894-897 (1962)
Dehydrochlorination of poly (vinyl chloride) by aqueous sodium hydroxide solution under two-phase conditions.
Kise H.
Journal of Polymer Science, 20(11), 3189-3197 (1982)
Some physical properties of aqueous solutions of tetramethylammonium bromide and tetramethylammonium iodide
Levien BJ.
Australian Journal of Chemistry, 18(8), 1161-1170 (1965)
Kinetic study of the base-catalyzed hydrolysis of novel high-spin hydrophobic Fe(II)-azomethine amino acid chelates: Salt and structure effects on the reactivity
Russian Journal of General Chemistry, 85, 168-172 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service