Skip to Content
Merck
All Photos(1)

Key Documents

181641

Supelco

Poly(Bisphenol A carbonate)

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

mol wt

average Mn 17,000 (typical)
average Mw 29,000 (typical)

technique(s)

gel permeation chromatography (GPC): suitable

density

1.2 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC(C)(c1ccc(O)cc1)c2ccc(O)cc2

InChI

1S/C15H16O2.CH2O3/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(17)10-6-12;2-1(3)4/h3-10,16-17H,1-2H3;(H2,2,3,4)

InChI key

XSXWYGABGYBZRM-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

General description

Poly(Bisphenol A carbonate) (PC) is a thermoplastic material. It is injected in molding operations and is majorly studied to understand its thermal behaviour, especially thermal degradation property. It can be synthesized by inducing supercritical CO2 to crystallinity in low molecular weight polycarbonate beads using solid-state polymerization method.

Application

It was used to study the lipase degradation and monitored using gel permeation chromatography (GPC).

Analysis Note

A lot specific analysis is included with each unit

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Rodan-Legrain et al.
Nature nanotechnology, 16(7), 769-775 (2021-05-05)
Magic-angle twisted bilayer graphene (MATBG) has recently emerged as a highly tunable two-dimensional material platform exhibiting a wide range of phases, such as metal, insulator and superconductor states. Local electrostatic control over these phases may enable the creation of versatile
Crystallization and solid-state polymerization of poly (bisphenol A carbonate) facilitated by supercritical CO2.
Gross, Stephen M., et al.
Macromolecules, 33.1, 40-45 (2000)
Dynamics of lipase catalyzed enzymatic degradation of poly (bisphenol-A carbonate).
Sivalingam, G., and Giridhar Madras
Journal of Applied Polymer Science, 91.4, 2391-2396 (2004)
Hiroshi Yabu et al.
Langmuir : the ACS journal of surfaces and colloids, 22(11), 4992-4997 (2006-05-17)
We describe here a preparation of pincushion structures with holes, hexagonally arranged microrings, and microdots by simple heating and peeling of self-organized honeycomb-patterned films. We have reported that the honeycomb-patterned films can be prepared by casting the solution of an
Vianney Souplet et al.
Methods in molecular biology (Clifton, N.J.), 570, 287-297 (2009-08-04)
Microarrays are frequently prepared on microscope glass slides. However, glass substrates can break or cut and thus can lead to the contamination of the manipulator during the analysis of biological samples. Alternately, bisphenol A polycarbonate (PC) is shock-resistant and, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service