Skip to Content
Merck
All Photos(1)

Key Documents

15821

Millipore

Klebsiella Selective Supplement

suitable for microbiology

Synonym(s):

Carbenicillin, α-Carboxybenzylpenicillin solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2O6S
Molecular Weight:
378.40
EC Number:
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Klebsiella spp.

SMILES string

CC1(C)S[C@@H]2[C@H](NC(=O)C(C(O)=O)c3ccccc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C17H18N2O6S/c1-17(2)11(16(24)25)19-13(21)10(14(19)26-17)18-12(20)9(15(22)23)8-6-4-3-5-7-8/h3-7,9-11,14H,1-2H3,(H,18,20)(H,22,23)(H,24,25)/t9?,10-,11+,14-/m1/s1

InChI key

FPPNZSSZRUTDAP-UWFZAAFLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For selective isolation and detection of Klebsiella species.

Biochem/physiol Actions

Carbenicillin is an ampicillin analog with structure related to benzyl penicillin. It is effective against Gram-negative bacteria and exhibits limited activity against Gram-positive bacteria. Carbenicillin binds and inhibits the enzymes involved in the synthesis of bacterial cell wall. Since carbenicillin is less sensitive to β-lactamases than ampicillin, it is effective against ampicillin-resistant bacteria. It has superior stability at low pH and acts as a selection agent. Effective concentration of carbenicillin: 50 to 100 μg/ml.

Components

(per vial, sufficient for 500 ml medium)
Carbenicillin 25 mg

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Emily K Davenport et al.
Antimicrobial agents and chemotherapy, 58(8), 4755-4761 (2014-06-11)
We investigated biofilms of two pathogens, Acinetobacter baumannii and Staphylococcus aureus, to characterize mechanisms by which the extracellular polymeric substance (EPS) found in biofilms can protect bacteria against tobramycin exposure. To do so, it is critical to study EPS-antibiotic interactions
Ilenys M Pérez-Díaz et al.
Food microbiology, 77, 10-20 (2018-10-10)
Limited documentation of the cucumber fermentation microbiome has impeded the understanding of the role of microbes on the quality of finished products. We characterized the microbiome of fresh and fermented cucumber samples using culture dependent and independent techniques, with an
Pingfeng Zhang et al.
PloS one, 9(7), e103518-e103518 (2014-07-25)
The HSP70 family of molecular chaperones function to maintain protein quality control and homeostasis. The major stress-induced form, HSP70 (also called HSP72 or HSPA1A) is considered an important anti-cancer drug target because it is constitutively overexpressed in a number of
Paul Yenerall et al.
Cell chemical biology, 27(1), 105-121 (2019-12-31)
RUVBL1 and RUVBL2 (collectively RUVBL1/2) are essential AAA+ ATPases that function as co-chaperones and have been implicated in cancer. Here we investigated the molecular and phenotypic role of RUVBL1/2 ATPase activity in non-small cell lung cancer (NSCLC). We find that RUVBL1/2
Gili Hrusa et al.
Applied and environmental microbiology, 81(8), 2900-2909 (2015-02-15)
Sodalis glossinidius is an intra- and extracellular symbiont of the tsetse fly (Glossina sp.), which feeds exclusively on vertebrate blood. S. glossinidius resides in a wide variety of tsetse tissues and may encounter environments that differ dramatically in iron content.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service