Skip to Content
Merck
All Photos(1)

Key Documents

03970590

Epigallocatechin gallate

primary reference standard

Synonym(s):

(−)-Epigallocatechin gallate, (−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-gallate, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxy-flavane-3-gallate, EGCG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18O11
CAS Number:
Molecular Weight:
458.37
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

storage temp.

−20°C

SMILES string

Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)c4cc(O)c(O)c(O)c4

InChI

1S/C22H18O11/c23-10-5-12(24)11-7-18(33-22(31)9-3-15(27)20(30)16(28)4-9)21(32-17(11)6-10)8-1-13(25)19(29)14(26)2-8/h1-6,18,21,23-30H,7H2/t18-,21-/m1/s1

InChI key

WMBWREPUVVBILR-WIYYLYMNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ichihashi et al.
Journal of dermatological science, 23 Suppl 1, S45-S50 (2000-04-15)
Reactive oxygen species (ROS) have been shown to be responsible for inducing DNA damage after ultraviolet radiation (UV). Antioxidant, vitamin E and epigallocatechin gallate extracted from green tea, applied topically to the skin, delayed the onset of UV-induced skin cancer
Masaki Kanadzu et al.
Cancer letters, 241(2), 250-255 (2005-11-24)
(-)-Epigallocatechin gallate (EGCG), a catechin polyphenol component, is the main ingredient of green tea extract. Recently, increasing attention has been given to its anti-oxidant effects. However, several studies reported the oxidative effects of EGCG, suggesting that EGCG had a dual
Xiaofeng Meng et al.
Chemical research in toxicology, 15(8), 1042-1050 (2002-08-20)
(-)-Epigallocatechin gallate (EGCG), the most abundant tea catechin, has been proposed to be beneficial to human health based on its strong antioxidative and other biological activities in vitro. Inadequate knowledge regarding the bioavailability and biotransformation of EGCG in humans, however
Olivier M Dorchies et al.
American journal of physiology. Cell physiology, 290(2), C616-C625 (2006-01-13)
Duchenne muscular dystrophy is a frequent muscular disorder caused by mutations in the gene encoding dystrophin, a cytoskeletal protein that contributes to the stabilization of muscle fiber membrane during muscle activity. Affected individuals show progressive muscle wasting that generally causes
Silvia A Mandel et al.
Journal of Alzheimer's disease : JAD, 15(2), 211-222 (2008-10-28)
Although much progress has been made in understanding the pathogenesis of Alzheimer's disease (AD), the current therapeutic approaches are merely symptomatic, intended for the treatment of cognitive symptoms, such as disturbances in memory and perception. Novel promising strategies suggest the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service