Skip to Content
Merck
All Photos(1)

Documents

00720585

Apigenin 7-glucoside

primary reference standard

Synonym(s):

4′,5,7-Trihydroxyflavone 7-glucoside, Apigenin-7-O-glucoside, Apigetrin, Cosmosiin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
65669
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O10/c22-8-16-18(26)19(27)20(28)21(31-16)29-11-5-12(24)17-13(25)7-14(30-15(17)6-11)9-1-3-10(23)4-2-9/h1-7,16,18-24,26-28H,8H2/t16-,18-,19+,20-,21-/m1/s1

InChI key

KMOUJOKENFFTPU-QNDFHXLGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: achillea ginkgo humulus hypericum melissa mentha pimpinella plantago salvia silybum thymus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sharaf
Natural product research, 22(18), 1620-1623 (2008-12-17)
The ethanolic extract of the seeds of Trifolium alexandrinum afforded a new naturally occurring compound identified as methyl-alpha-glucose. The known compounds quercetin, kaempferol, apigenin 7-O-beta-D-glucoside and the nucleoside xanthosine were also isolated. The isolated compounds were identified by spectroscopic (UV
Soninkhishig Tsolmon et al.
Molecular nutrition & food research, 55 Suppl 1, S93-S102 (2011-04-12)
Induction of cancer-cell differentiation is an alternative approach for cancer chemotherapy. There are numerous studies that diets containing an abundance of fruits and vegetables have protection against cancers, and the main agents thought to provide such protective effect are flavonoids.
Mostafa Hojati et al.
Journal of plant physiology, 168(8), 782-791 (2011-01-07)
In this experiment, the possibility of enhancing the water deficit stress tolerance of chamomile (Matricaria chamomilla L.) during two growth stages by the exogenous application of hexaconazole (HEX) was investigated. To improve water deficit tolerance, HEX was applied in three
D I Pisarev et al.
Bulletin of experimental biology and medicine, 150(6), 714-717 (2012-01-12)
Juniperus oblonga Bieb is widely spread in the Caucasus Mountains, particularly in its eastern and southern regions. Diuretic effect of juniper berries is determined by the presence of volatile oils and polyphenol complex, particularly flavonoids. Flavonoids were extracted from raw
Kazutoshi Shindo et al.
Journal of natural medicines, 62(2), 247-248 (2008-04-12)
The edible flower of Torenia fournieri Linden ex E. Fourn was found to possess potent antioxidative activity in a rat brain homogenate model. Bioassay-guided isolation of the active compounds from a CH(2)Cl(2)-MeOH (1:1) extract led to the isolation of acteoside

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service