Skip to Content
Merck
All Photos(1)

Key Documents

WBULP-100ML

Millipore

Immobilon® ECL UltraPlus Western HRP Substrate

Synonym(s):

HRP substrate, chemiluminescent detection substrate, detection substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105332
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

Immobilon® ECL UltraPlus Western HRP Substrate - 100mL,

product line

Immobilon®

Quality Level

technique(s)

dot blot: suitable
immunoblotting: suitable
western blot (chemiluminescent): suitable

Components

  • 50mL Enhancer Solution
  • 50mL Peroxide Solution

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service