Skip to Content
Merck
All Photos(1)

Key Documents

SIMPAKKD2

Milli-Q

Kit: SimPak® 2 Purification

For Simplicity® systems with Elix® / RO / distilled water feed

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.13

Quality Level

packaging

pkg of 1 kit

compatibility

for use with Simplicity®

shipped in

ambient

General description

The SimPak® 2 Purification kit contains one each Purification Pack, SimFilter, and 0.05 μm Reservoir Vent Filter. SimPak® 2 Purification kit for use with Simplicity® systems with Elix® / RO / distilled water feed.

Legal Information

ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPAK is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service