SCT137
BioTracker 633 Red Mitochondria Dye
Live cell imaging mitochondrial dye that stains the membrane of mitochondria used to detect cell viability, metabolic activity and overall cell health.
Synonym(s):
Live cell imaging probe
About This Item
Recommended Products
technique(s)
cell based assay: suitable
detection method
fluorometric
shipped in
ambient
General description
BioTracker Mitochondria dyes are fluorogenic stains for staining mitochondria in live cells. The dyes are membrane permeable and become brightly fluorescent upon accumulation in the mitochondrial membrane. Staining is lost when mitochondria become depolarized during cell death, allowing monitoring of cell viability.
The BioTracker 633 Red Mitochondria Dye can be detected using the red Cy5 channel. The fluorescence is dependent on mitochondrial membrane potential, and the dye can be used to monitor changes in mitochondrial membrane potential in cells during apoptosis.
Spectral Properties
Absorbance: 622nm
Emission: 648nm
Application
Cell Imaging
Live Cell Dye
Quality
Absorbance: 622nm
Emission: 648nm
Physical form
Disclaimer
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service