Skip to Content
Merck
All Photos(1)

Key Documents

SCR031

Sigma-Aldrich

Cryopreserved Mouse Spinal Cord Neural Stem Cells

The Cryopreserved Mouse Spinal Cord Neural Stem Cells provides ready-to use primary neural stem cells & is isolated from the spinal cord of embryonic day 15-18 (E15-E18) C57/BL6 mice.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
eCl@ss:
32011203
NACRES:
NA.81

biological source

mouse

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable

input

sample type neural stem cell(s)

shipped in

liquid nitrogen

General description

Millipore′s Cryopreserved Mouse Spinal Cord Neural Stem Cells provides ready-to use primary neural stem cells isolated from the spinal cord of embryonic day 15-18 (E15-E18) C57/BL6 mice. These primary neural stem cells may be used for a variety of research applications including drug development, for studies of neurotoxicity, neurogenesis, electrophysiology, neurotransmitter and receptor functions and CNS diseases and disorders.

We recommend that Millipore′s Cryopreserved Mouse Spinal Cord Neural Stem Cells (Catalog No. SCR031) be used in conjunction with the Neural Stem Cell Marker Characterization Kit (Catalog No. SCR019) and differentiation assays that demonstrate multipotentiality of the starting cell population.

For Research Use Only; not for use in diagnostic procedure.

Cell Line Description

Neural Stem Cells

Application

Research Category
Stem Cell Research

Packaging

1 x 106 cells/vial

Components

1x10e6 viable Mouse Spinal Cord Neural Stem Cells: (Part No. 2004116) derived from mixed gender C57/BL6 embryonic day 15-18 mice, cryopreserved. Store in liquid nitrogen.

Storage and Stability

When stored at the recommended storage conditions (liquid nitrogen), mouse spinal cord neural stem cells are stable up to the expiration date. Do not expose to elevated temperatures. Discard any remaining reagents after the expiration date. We recommend that the cells be used within ten passages.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
cOmplete is a trademark of Roche

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service