MXHAWG124
Milliflex® funnel
100mL, 0.45μm white gridded for bioburden and water analysis with the Milliflex® system
About This Item
Recommended Products
material
gridded filter
mixed cellulose esters (MCE)
polypropylene
styrene-acrylonitrile (SAN)
white filter
Quality Level
sterility
sterile; ethylene oxide treated
manufacturer/tradename
Milliflex®
parameter
100 mL sample volume
filter diam.
51 mm
funnel capacity
100 mL
matrix
MF-Millipore™
pore size
0.45 μm pore size
input
sample type cosmetics
sample type beer
sample type pharmaceutical(s)
sample type bottled water
application(s)
bioburden testing
cosmetics
pharmaceutical
water monitoring
shipped in
ambient
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service