Skip to Content

MAC148

Anti-Cas9 Antibody, clone 8G4

Synonym(s):

CRISPR-associated endonuclease Cas9/Csn1, SpCas9, SpyCas9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
8G4, monoclonal
Application:
IF, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

8G4, monoclonal

mol wt

calculated mol wt 158.44 kDa, observed mol wt ~210 kDa

purified by

using protein G

species reactivity

all

packaging

antibody small pack of 100

technique(s)

immunofluorescence: suitable, western blot: suitable

isotype

IgG2aλ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

Streptococcus pyogenes ... Cas9(69900935)

General description

CRISPR-associated endonuclease Cas9/Csn1 (UniProt: Q99ZW2; also known as SpCas9, SpyCas9, Cas9) is encoded by the Cas9 gene (Gene ID: 69900935) in Streptococcus pyogenes. CRISPR (clustered regularly interspaced short palindromic repeat) is an adaptive immune system that provides protection against mobile genetic elements (viruses, transposable elements, and conjugative plasmids). CRISPR clusters contain spacers, sequences complementary to antecedent mobile elements, and target invading nucleic acids. Cas9 has a bi-lobed architecture with a recognition lobe (residues 41-425) and a discontinuous nuclease lobe (NUC, residues 1-40 and 453-1053); the crRNA-target DNA lies in a channel between the 2 lobes. The NUC lobe has 2 endonuclease domains. The discontinuous RuvC-like domain in NUC cleaves the target DNA non-complementary to crRNA while the HNH nuclease domain cleaves the target DNA complementary to crRNA. Cas9 serves as a RNA-guided DNA endonuclease that cleaves double-stranded DNA targets with a protospacer adjacent motif (PAM) and complementarity to the guide RNA. (Ref.: Fonfara, I., et al. (2014). Nucleic Acids Res. 42(4); 2577-2590; Jinek, M., et al. (2012). Science. 337(6096); 816-821).

Immunogen

His-tagged, full-length, recombinant Cas9 from Streptococcus pyogenes.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from HeLa cells overexpressing Cas9.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Cas9 in lysate from HeLa cells overexpressing Cas9.

Tested Applications

Immunofluorescence Analysis: A 1:500 dilution from a representative lot detected Cas9 in Cell line stably expressing Cas9-NLS (A) versus wild type parental cell line (B) (Courtesy of Germán Camargo Ortega (Institute of Stem Cell Research, Helmholtz Munich,. Germany).

Western Blotting Analysis: A representative lot detected Cas9 in Western Blotting application. (Camargo Ortega, G., et al. (2019). Nature. 567(7746):113-117).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 8G4 is a rat monoclonal antibody that specifically detects Cas9.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library