Skip to Content
Merck
All Photos(2)

Documents

MABS87

Sigma-Aldrich

Anti-GPR177 Antibody, clone YJ5

clone YJ5, from mouse

Synonym(s):

Protein wntless homolog, Integral membrane protein GPR177, Protein evenness interrupted homolog, Putative NF-kappa-B-activating protein 373, WLS, EVI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

YJ5, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WLS(79971)

General description

Gpr177 (WLS, EVI), the mouse ortholog of Drosophila WIs, is expressed during mouse embryogenesis. It is essential for the patterning of the anterior-posterior axis during mammalian development and is activated by â-catenin and LEF/TCF-dependent transcription. Upon GPR177 activation, Wnt binds with and modifies the cellular distribution of GPR177, leading to a feedback regulatory mechanism involving Wnt expression and signaling.

Immunogen

Histidine-tagged recombinant protein corresponding to human GPR177.

Application

Anti-GPR177 Antibody, clone YJ5 is an antibody against GPR177 for use in WB & IC.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected GPR177 in HeLa cells (Prof. D.M. Virshup, Program in Cancer and Stem Cell Biology, Duke-NUS Graduate Medical School.) (Also reference Coombs, G.S., et al. (2010). J Cell Sci. 123(19):3357-3367).
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:5,000 dilution of this antibody detected GPR177 in 10 µg of HeLa cell lysate.

Target description

~62 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induction and transport of Wnt 5a during macrophage-induced malignant invasion is mediated by two types of extracellular vesicles.
Menck, K; Klemm, F; Gross, JC; Pukrop, T; Wenzel, D; Binder, C
Oncotarget null
Wenhui Wang et al.
Neoplasia (New York, N.Y.), 18(12), 711-723 (2016-11-17)
Aberrant activation of Wnt/β-catenin signaling plays a key role in the onset and development of hepatocellular carcinomas (HCC), with about half of them acquiring mutations in either CTNNB1 or AXIN1. However, it remains unclear whether these mutations impose sufficient β-catenin
Leandro S D'Abronzo et al.
American journal of clinical and experimental urology, 10(5), 299-310 (2022-11-01)
Resistance to androgen receptor (AR) targeted therapies remains as the main reason for most prostate cancer related deaths. Lineage plasticity resulting in altered, treatment insensitive prostate tumor cell phenotypes such neuroendocrine differentiated prostate cancer is a common manifestation within resistant
Alan P Lombard et al.
American journal of clinical and experimental urology, 7(4), 203-214 (2019-09-13)
De-regulation of Wnt signaling pathways has been shown to be associated with progression of castration-resistant prostate cancer and more recently, studies indicate that both canonical and non-canonical Wnt pathways may mediate resistance to anti-androgen therapies such as enzalutamide. However, the
Jonathan Stewart et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 28(3), 428-436 (2014-09-27)
The oncogenic role of WNT is well characterized. Wntless (WLS) (also known as GPR177, or Evi), a key modulator of WNT protein secretion, was recently found to be highly overexpressed in malignant astrocytomas. We hypothesized that this molecule may be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service