Skip to Content
Merck
All Photos(1)

Key Documents

MABN380

Sigma-Aldrich

Anti-APP-C99 Antibody, clone mC99(70–80)

ascites fluid, clone mC99(70-80), from mouse

Synonym(s):

Amyloid beta A4 protein, ABPP, APPI, APP, Alzheimer disease amyloid protein, Cerebral vascular amyloid peptide, CVAP, PreA4, Protease nexin-II, PN-II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
mC99(70-80), monoclonal
application:
IF
IHC
IP
WB
species reactivity:
human, mouse
technique(s):
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
10

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

mC99(70-80), monoclonal

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

General description

Amyloid Precursor Protein (APP) is expressed in the brain, kidney, heart, and spleen of fetal tissues; it is induced during neuronal differentiation. The most-substantiated role for APP is in synaptic formation and repair; its expression is upregulated during neuronal differentiation and after neural injury. In adult brain, highest expression of APP is found in the frontal lobe of the cortex. Moderate expression is observed in the cerebellar cortex, the posterior perisylvian cortex-opercular gyri and the temporal associated cortex. Defects in APP are a cause of autosomal dominant Alzheimer′s disease (AD). UP on intracerebroventricular injection of this antibody, A-beta levels significantly decreased in the brain of APPPS1 mouse model of AD. A related antibody, Cat. No. MABN381, Anti-APP-C99, clone mC99(1-7) was raised against a slightly different region of the same APP-C99 protein.

Immunogen

Recombinant protein corresponding to Human APP-C99.

Application

Anti-APP-C99 Antibody, clone mC99(70–80) is a highly specific mouse monoclonal antibody, that targets APP & has been tested in western blotting, IP, IHC & IF.
Immunoprecipitation Analysis: A representative lot immunoprecipitated APP-C99 in HEK APPSwe cell lysate (Houacine, J., et al. (2012). Neurobiol Aging. 33(11):2704-2714.).

Immunohistochemistry Analysis: A representative lot detected APP-C99 in APPS1 mice and in human Alzheimer brain tissues
(Houacine, J., et al. (2012). Neurobiol Aging. 33(11):2704-2714.).

Immunofluoresence Analysis: A representative lot detected APP-C99 in HEK APPSwe cells (Houacine, J., et al. (2012). Neurobiol Aging. 33(11):2704-2714.).
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in DAPT treated HEK293 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected APP-C99 in 10 µg of DAPT treated HEK293 cell lysate.

Target description

~95 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Mouse monoclonal IgGκ ascites
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You-Chang Oh et al.
Nutrients, 12(7) (2020-07-11)
Banhasasim-tang (BHS) is an herbal medicine that has been widely used in East Asia to treat various symptoms associated with upper abdomen swelling. BHS has not been studied previously for neuroinflammation or cognitive disorder. Here, we use a lipopolysaccharide (LPS)
Hana Na et al.
Neuropharmacology, 168, 108017-108017 (2020-03-03)
Calcitonin gene-related peptide (cGRP) receptor antagonists effectively treat migraine through reducing neuroinflammation, vasoconstriction and possibly neruogenesis. Since neuroinflammation is also involved in the pathogenesis of Alzheimer's diseases (AD), we hypothesized and tested if a cGRP receptor antagonist, BIBN 4096 BS
Qian Guo et al.
Acta pharmaceutica Sinica. B, 9(3), 590-603 (2019-06-14)
Gene therapy represents a promising treatment for the Alzheimer׳s disease (AD). However, gene delivery specific to brain lesions through systemic administration remains big challenge. In our previous work, we have developed an siRNA nanocomplex able to be specifically delivered to
Hana Na et al.
American journal of Alzheimer's disease and other dementias, 36, 15333175211012867-15333175211012867 (2021-06-18)
Intraperitoneal injection of amylin or its analog reduces Alzheimer's disease (AD) pathology in the brains. However, self-injecting amylin analogs is difficult for patients due to cognitive deficits. This work aims to study the effects of amylin on the brain could
Haruka Okabayashi et al.
Biological & pharmaceutical bulletin, 47(1), 192-195 (2024-01-18)
Plasmalogens are a family of glycerophospholipids containing one vinyl-ether bond at the sn-1 position in the glycerol backbone, and play important roles in cellular homeostasis including neural transmission. Therefore, reductions of plasmalogens have been associated with neurodegenerative disorders, such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service