Skip to Content
Merck
All Photos(1)

Key Documents

MAB5556

Sigma-Aldrich

Anti-Nicastrin Antibody

ascites fluid, clone 9C3, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
9C3, monoclonal
application:
ICC
IP
WB
species reactivity:
rat, human, mouse
technique(s):
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
9

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

9C3, monoclonal

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NCSTN(23385)

Specificity

Nicastrin.

Immunogen

Synthetic peptide corresponding to the C terminal 18 a.a. of mouse Nicastrin. Due to sequence similarity, it is expected that the antibody will react with most mammals.

Application

Anti-Nicastrin Antibody is an antibody against Nicastrin for use in IP, WB & IC.
Western blot. The antibody recognizes a doublet of ~100-110 kDa. Suggested antibody dilution buffer is TBS with 0.1% Tween-20 and 5% non-fat milk powder. Suggested incubation time is overnight at 2-8°C or 1-2 hours at room temperature.

Immunocytochemistry

Immunoprecipitation. Suggested tissue/cell lysis buffer is 1% Triton X100 or 2% CHAPS. Suggested final reaction volume is 1000 μL with a final protein concentration in the reaction mix of 1 mg/mL. Suggested incubation time is overnight at 2-8°C with rotating. The antibody is known to co-precipitate in CHAPS: presenilin, Aph-1 and Pen-2. Optimal working dilutions must be determined by end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolanta L Lundgren et al.
Journal of neurochemistry, 135(3), 606-615 (2015-08-25)
Synaptic degeneration and accumulation of the neurotoxic amyloid β-peptide (Aβ) in the brain are hallmarks of Alzheimer disease. Aβ is produced by sequential cleavage of the amyloid precursor protein (APP), by the β-secretase β-site APP cleaving enzyme 1 (BACE1) and
Mitsuhiro Inoue et al.
The FEBS journal, 282(14), 2587-2599 (2015-04-22)
The transmembrane protease complex γ-secretase is a key enzyme in Alzheimer disease pathogenesis as it liberates the neurotoxic amyloid β-peptide (Aβ); however, the mechanism of regulation of its activity in various cell types and subcellular compartments is largely unknown. Several
Yasuhiro Teranishi et al.
The FEBS journal, 282(17), 3438-3451 (2015-06-23)
γ-Secretase is a transmembrane protease complex that is responsible for the processing of a multitude of type 1 transmembrane proteins, including the amyloid precursor protein and Notch. γ-Secretase processing of amyloid precursor protein results in the release of the amyloid
Rocío Pérez-González et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(9), 12922-12931 (2020-08-11)
Pleiotropic roles are proposed for brain extracellular vesicles (EVs) in the development of Alzheimer's disease (AD). Our previous studies have suggested a beneficial role for EVs in AD, where the endosomal system in vulnerable neurons is compromised, contributing to the
gamma-Secretase dependent production of intracellular domains is reduced in adult compared to embryonic rat brain membranes.
Fr?nberg, J; Karlstrom, H; Winblad, B; Tjernberg, LO; Frykman, S
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service