Skip to Content
Merck
All Photos(1)

Documents

MAB5272

Sigma-Aldrich

Anti-Neural Cell Adhesion Molecule L1 Antibody, clone 324

clone 324, Chemicon®, from rat

Synonym(s):

CD171, N-CAM L1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

324, monoclonal

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Families of adhesion molecules which share common carbohydrate domains do exist, despite the structural and functional diversity of these glycoproteins. These include the Ca2+-independent neural adhesion molecules: N-CAM, myelin associated glycoprotein (MAG) and L1. L1 is involved in neuron-neuron adhesion, neurite fasciculation, outgrowth of neurites, cerebellar granule cell migration, neurite outgrowth on Schwann cells and interactions among epithelial cells of intestinal crypts

Specificity

Monoclonal antibody against neural cell adhesion molecule L1 from rat-rat hybrid cells.

Immunogen

Rats were immunized with a glycoprotein fraction from cerebellum of 8-10 day old C57BL/6J mice.

Application

This Anti-Neural Cell Adhesion Molecule L1 Antibody, clone 324 is validated for use in IC, IH, WB for the detection of Neural Cell Adhesion Molecule L1.
Western blotting:
20 μg/mL was used on a previous lot.

Immunocytochemistry:
5-10 μg/mL was used on a previous lot. Cells must be fixed with methanol for 10 minutes at -20°C in the middle log phase.

Immunohistochemistry: Clone 324 is sensitive to fixation. Fresh frozen, acetone or -20C methanol fixed tissues are recommended. Traditional 4% PFA typically does not work well. Other fixatives have not been tested.

Optimal working dilutions must be determined by end user.

Target description

120-140 kDa

Physical form

Format: Purified
Rat monoclonal in buffer containing 0.02 M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% sodium azide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuron glia-related cell adhesion molecule (NrCAM) promotes topographic retinocollicular mapping.
Dai, J; Buhusi, M; Demyanenko, GP; Brennaman, LH; Hruska, M; Dalva, MB; Maness, PF
Testing null
ADAM17 is critical for multipolar exit and radial migration of neuronal intermediate progenitor cells in mice cerebral cortex.
Li, Q; Zhang, Z; Li, Z; Zhou, M; Liu, B; Pan, L; Ma, Z; Zheng, Y
Testing null
Evidence that descending cortical axons are essential for thalamocortical axons to cross the pallial-subpallial boundary in the embryonic forebrain.
Chen, Y; Magnani, D; Theil, T; Pratt, T; Price, DJ
Testing null
Yuhki Saito et al.
eLife, 5 (2016-05-26)
The neuron specific RNA-binding proteins NOVA1 and NOVA2 are highly homologous alternative splicing regulators. NOVA proteins regulate at least 700 alternative splicing events in vivo, yet relatively little is known about the biologic consequences of NOVA action and in particular
Morgane Sonia Thion et al.
Cell, 172(3), 500-516 (2017-12-26)
Microglia are embryonically seeded macrophages that contribute to brain development, homeostasis, and pathologies. It is thus essential to decipher how microglial properties are temporally regulated by intrinsic and extrinsic factors, such as sexual identity and the microbiome. Here, we found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service