Skip to Content
Merck
All Photos(1)

Key Documents

MAB16985B

Sigma-Aldrich

Anti-MCAM Antibody, clone P1H12, biotin conjugated

clone P1H12, Chemicon®, from mouse

Synonym(s):

MUC18, CD146

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1H12, monoclonal

species reactivity

mouse, human, canine

should not react with

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MCAM(4162)

General description

MUC18 antigen (CD146), a member of the immuoglobulin superfamily, is an intrinsic membrane glycoprotein of 110-120 kDa found on the surface of endothelial cells, bone marrow fibroblasts and various melanomas. MUC18 has been used as a marker of tumor progression in human melanoma because expression in those tumors correlates strongly with poor prognosis and the development of metastic disease. In addition, a number of human T, B and myeloid leukemic cell lines seem to express MUC18. The close structural relationship with N-CAM and related molecules suggests that MUC18 may be also a developmentally regulated cell adhesion molecule (Melanoma adhesion molecule or MCAM).

Specificity

Detects circulating endothelial cells in human. Expected to cross-react with dog & mouse.
P1H12 reacts specifically with CD146 {MCAM, MUC18}. In blood and marrow P1H12 reacts only with endothelial cells, and has been used to detect circulating endothelial cells in human, dogs and mouse samples. It positively stains normal, primary HUVEC and MVEC cultures and the endothelial cells of all vessels in normal frozen sections of human skin, intestine, ovary tonsil, lymph node, lung, and kidney. It does not stain carcinoma cell lines HT-29 and COLO205, and M21, the T cell lines Jurkat and HuT78, fibroblasts, HL-60 or CHO cells, or EBV-transformed B cell lines, although it expected that CD146 expression is present in many tumor lines. P1H12 does not stain normal monocytes, granulocytes, red cells, platelets, T cells or B cells from marrow or peripheral blood; nor does it detect marrow megakaryocytes or the megakaryoblast line HU3. The peripheral blood cells that do stain with P1H12 also are positive for both von Willebrand Factor (vWF) and thrombomodulin (the combined expression of which is limited to endothelium), and they stain for flt and flk. In recent testing, melanoma A2058, SKMEL.3 and A375.S2 are positive, while murine melanoma M3 is negative. One smooth muscle line (HISM) was positive and one negative (TIGHA-VSMC). Human blood lymphoctes are negative with or without prior stimulation.

Immunogen

Immunization of mice with HUVEC

Application

Immunocytochemistry: 1-10 μg/mL. Works best on EDTA or Trypsin lifted endothelial cells.

Immunohistochemistry: 1-10 μg/mL. 4% PFA for 30min RT or <2hrs @4°C. Block w/ 1%BSA/0.2%tween20/PBC for 30min. Works well in frozen tissue; fixed or unfixed.

Immunoprecipitation: 1-10 μg/mL

ELISA: 1-10 μg/mL

Facs Analysis: 1-10 μg/mL

Optimal working dilutions and protocols must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-MCAM Antibody, clone P1H12, biotin conjugated is validated for use in ELISA, FC, IP, IC, IH for the detection of MCAM.

Physical form

Purified immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl, pH=7.6, 15 mg/nL BSA containing 0.1% sodium azide as a preservative.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Memet Emin et al.
Science translational medicine, 8(320), 320ra1-320ra1 (2016-01-08)
Obstructive sleep apnea (OSA), characterized by intermittent hypoxia (IH) during transient cessation of breathing, triples the risk for cardiovascular diseases. We used a phage display peptide library as an unbiased approach to investigate whether IH, which is specific to OSA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service