Skip to Content
Merck
All Photos(1)

Documents

MAB1569

Sigma-Aldrich

Anti-Microglia Antibody, clone 25F9

clone 25F9, Chemicon®, from mouse

Synonym(s):

Macrophages, Late State Inflammatory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

25F9, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

General description

ANTIGEN DISTRIBUTION:

Isolated cells: Absent on freshly isolated monocytes and other blood cells; present on 40-50% of human monocytes after 6-7 day culture, also positive on some melanoma and carcinoma cell lines. Tissue sections: Kupffer cells, histiocytes (skin), macrophages of the thymus, in the germinal centers of lymph nodes and spleen, in mamma carcinoma, melanoma, osteocarcinoma and gastric cancer; excema, sarcoidosis, BCG granuloma; synovial lining cells, tuberculoid leprosy; no expression in lepramatous leprosy.

Specificity

Human late stage inflammatory macrophages. The monoclonal is suitable for use on BAL (bronchial lavage fluids) and other lavages. The antigen recognized by MAB1569 is an 86 kDa protein (unreduced conditions), probably a glycoprotein on the cell surface and within the cytoplasm of mature macrophages. It is stable to formaldehyde fixation and paraffin embedding. Enzyme digestion is recommended. The antibody has also been reported to recognize microglia stem cells (X International Congress of Neuropathology, Stockholm, le 11 September,1986.PosterA-701).

Antigen distribution: absent from freshly isolated monocytes and other blood cells; present on 40-50% of human monocytes after 6-7 days in culture, also positive on some melanoma and carcinoma lines. In tissue sections, clone identifies Kupffer cells, histiocytes (skin), macrophages of the thymus, in te germinal centers of lymph nodes and spleen, in mamma carcinoma, melanoma, osteocarcinoma and gastric cancer; excema, sarcoidosis, BCG granuloma;synovial lining cells, tuberculoid leprosy, however no expression in lepramatous leprosy.

Species reactivity is seen in human, rhesus monkey, and pig, other species not tested.

Application

Anti-Microglia Antibody, clone 25F9 detects level of Microglia & has been published & validated for use in FC, IC, IH.
Immunohistochemistry: 1:50-1:200 using human tonsil cryosection. The antibody is reactive on formalin fixed / paraffin sections. Enzyme digestion (proteinase pretreatement) is recommended.

Immunocytochemistry

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin from culture supernatant. Lyophilized from PBS, pH 7.2 containing 5 mg/mL BSA and 0.09% sodium azide. Reconstitute with 500 μL of sterile distilled water.

Storage and Stability

Maintain lyophilized material at 2-8°C for up to 6 months. After reconstitution maintain at -70°C for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Vogt et al.
EMBO molecular medicine, 8(1), 25-38 (2015-12-17)
Loss of plasticity-related gene 1 (PRG-1), which regulates synaptic phospholipid signaling, leads to hyperexcitability via increased glutamate release altering excitation/inhibition (E/I) balance in cortical networks. A recently reported SNP in prg-1 (R345T/mutPRG-1) affects ~5 million European and US citizens in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service