LSKP096
Plasmid Miniprep96 Kit
Plasmid & BAC Miniprep
Synonym(s):
Montage Plasmid Miniprep
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
Montage®
plate size
96 wells
application(s)
genomic analysis
shipped in
ambient
General description
Plasmid Miniprep96 Kit is a quick and easy-to-use kit that produces clean and reproducible DNA in half a time than traditional methods. Bind-wash-elute or centrifugation steps are not required. The kit is inclusive of all the reagents and disposable materials needed to purify plasmid DNA in a 96-well format. Purified samples are applicable for use in most of the sensitive downstream applications such as cloning, DNA sequencing, and transformation.
Application
Plasmid Miniprep96 Kit has been used to prepare DNA of the expression plasmids for automated apoptosis assay to detect activation of caspase 3.
Features and Benefits
- No centrifugation or precipitation steps required
- Minimum processing time
- Excellent purity, yields, and reproducibility
- Automation compatible
Components
includes 96-well filter plates and all the reagents and disposable materials you need to purify plasmid or BAC DNA
Physical form
Color Code: Clear
Legal Information
MONTAGE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
General Lab Use
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8A - Combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service