GX0185
Glycerol
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Synonym(s):
Glycerin, 1, 1,2,3-Propanetriol, Glycerin
About This Item
Recommended Products
grade
reagent
Quality Level
Agency
meets USP/NF
vapor density
3.1 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
698 °F
technique(s)
electrophoresis: suitable
impurities
≤0.003% Chlorinated compounds (as Cl)
≤0.05% Fatty acid esters (as butyric acid)
≤0.5% Water
≤2 ppm Heavy metals (as Pb)
ign. residue
≤0.005%
color
APHA: ≤10
refractive index
n20/D 1.474 (lit.)
pH
5 (20 °C, 100 g/L in H2O)
bp
182 °C/20 mmHg (lit.)
mp
20 °C (lit.)
transition temp
flash point 199 °C
density
1.25 g/mL (lit.)
anion traces
sulfate (SO42-): ≤0.001%
shipped in
ambient
storage temp.
room temp
SMILES string
OCC(O)CO
InChI
1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2
InChI key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Analysis Note
Assay (GC): 99.5% min
Chlorinated compounds (as Cl): 0.003% max
Color (APHA): 10 max
Fatty acid esters (as butyric acid): 0.05% max
Form: Clear liquid
Heavy metals (as Pb): 2ppm max
Infrared Spectrum: Conforms to standard
Neutrality: To pass test
Residue after ignition: 0.005% max
Substances darkened by sulfuric acid: To pass test
Sulfate (SO4): 0.001% max
Water (H2O): 0.5% max
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
390.2 °F - Pensky-Martens closed cup
Flash Point(C)
199 °C - Pensky-Martens closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service