Skip to Content
Merck
All Photos(1)

Key Documents

GSWP04700

Millipore

MCE Membrane Filter, 0.22 μm Pore Size

MF-Millipore, filter diam. 47 mm, hydrophilic, white

Synonym(s):

Cellulose nitrate membrane filter discs, MCE membrane filter, MF-Millipore Membrane Filter, 0.22 µm pore size, Mixed Cellulose Ester membrane filter discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

description

47 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

18 mL/min-cm2 water flow rate
2 L/min-cm2 air flow rate
75 °C max. temp.

filter diam.

47 mm

thickness

150 μm

gravimetric extractables

2%

color

white

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.22 μm pore size
75 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥3.52 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: GSWP

Application

  • Sterilizing filtration
  • Bioassays
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranjith Eswaran et al.
Marine environmental research, 159, 104997-104997 (2020-07-15)
The influence of changing salinity on community composition and functional activity (Bacterial Production (BP) and ectoenzyme activity) of major bacterial taxa was evaluated using microcosm experiments in a tropical monsoon influenced estuary. Natural bacterial inocula at different salinities, representing marine
Anna Alekseeva et al.
Carbohydrate polymers, 234, 115913-115913 (2020-02-20)
Rapid advances have been made in developing analytical technologies for characterization of highly heterogeneous active ingredients of complex drugs, such as pentosan polysulfate (PPS), active ingredient of the drug Elmiron®, approved by the Food and Drug Administration and marketed in

Questions

1–4 of 4 Questions  
  1. Can the PES filter be used to filter ethanol?

    1 answer
    1. This product is an MCE membrane, not a PES membrane. MCE is not recommended for use with ethanol. For a complete membrane chemical compatibility chart, please contact the local Technical Service office.

      Helpful?

  2. Should these filters be pre-wet before autoclaving or should they be prepared dry?

    1 answer
    1. This product does not need to be pre-wet. It should be removed from its packaging and wrapped in foil or any standard autoclave paper may be autoclaved at 121 °C for 15-20 minutes using the slow exhaust setting.

      Helpful?

  3. Hi, Do you have a filter that has both hydrophilic and hydrophobic properties?

    1 answer
    1. MCE Membrane Filter, 5.0 ?m Pore Size (SMEP0470W) is hydrophilic with 6 mm hydrophobic edge. Please refer to this link below for more information:
      https://www.sigmaaldrich.com/product/mm/smep0470w

      Helpful?

  4. Hello, can this filter be autoclaved before use? how would you recommend doing this?

    1 answer
    1. This item may be autoclaved at 121 °C for 15-20 minutes with slow exhaust.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service