FCMAB180F
Milli-Mark® Anti-CD13-FITC Antibody, clone WM-47
clone WM-47, Milli-Mark®, from mouse
Synonym(s):
Aminopeptidase N (AP-N), hAPN, Alanyl aminopeptidase, Microsomal aminopeptidase, Aminopeptidase M (AP-M), gp150, Myeloid plasma membrane glycoprotein CD13
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
FITC conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
WM-47, monoclonal
species reactivity
human
manufacturer/tradename
Milli-Mark®
technique(s)
flow cytometry: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... ANPEP(290)
Related Categories
General description
Anti-CD13, WM-47, was included in the Fourth and Fifth International Workshop and Conference on Leucocyte Differentiation Antigens and studies by a number of laboratories confirmed its reactivity with CD 13 (Ashmun, 1995; Look, 1989).
Specificity
Immunogen
Application
Inflammation & Immunology
Immunoglobulins & Immunology
Quality
Target description
Physical form
Storage and Stability
Analysis Note
PBMCs
Other Notes
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service