Skip to Content
Merck
All Photos(2)

Key Documents

ABE217

Sigma-Aldrich

Anti-CUX1 Antibody, a.a. 861

from rabbit, purified by affinity chromatography

Synonym(s):

Homeobox protein cut-like 1, CCAAT displacement protein, CDP, Homeobox protein cux-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CUX1(1523)

General description

Homeobox protein cut-like 1 (CUX1) is also known as CCAAT displacement protein (CDP) and Homeobox protein cux-1. CUX1 is thought to repress expression of developmentally regulated genes involved in mammalian development, likely through the prevention of CCAAT factor binding at promoters. CUX1 also represses T-cell receptor (TCR) beta enhancer function by binding to matrix attachment region beta (MARbeta), and is a component of the nf-munr repressor, which binds to matrix attachment regions (MARs) of the immunoglobulin heavy chain enhancer.

Specificity

Other homologies: Mouse (95% sequence homology).
This antibody recognizes the region of amino acids beginning at 861 of CUX1. This antibody is expected to cross react with all known isoforms of CUX1.

Immunogen

Epitope: Region of amino acids beginning at 861
GST-tagged recombinant protein corresponding to a region of amino acids beginning at 861 of human CUX1.

Application

Detect Cux1 using this rabbit polyclonal antibody, Anti-CUX1 Antibody, a.a. 861 validated for use in western blotting & IP.
Immunoprecipitation Analysis: 1.25 µg from a representative lot immunoprecipitated CUX1 in 500 µg of HeLa nuclear extract.

Alexa Fluor is a registered trademark of Life Technologies.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 0.25 µg/mL from a representative lot detected CUX1 in 10 µg of HeLa nuclear extract.

Target description

~200 kDa and ~110 kDa observed. Uniprot describes 11 different isoforms. A full length protein has been described at 200 kDa and is proteolytically cleaved into the p150, p110, p90, and p80 isoforms. Additionally, the p75 isoform is the result of alternative splicing (Sansregret, L, et al. (2008). Gene. 412(1-2):84-94.). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa nuclear extract

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nele Vollersen et al.
Bone research, 9(1), 48-48 (2021-11-12)
The recent identification of homozygous WNT1 mutations in individuals with osteogenesis imperfecta type XV (OI-XV) has suggested that WNT1 is a key ligand promoting the differentiation and function of bone-forming osteoblasts. Although such an influence was supported by subsequent studies
Balraj Singh et al.
PloS one, 11(7), e0159072-e0159072 (2016-07-09)
We have previously shown that only 0.01% cells survive a metabolic challenge involving lack of glutamine in culture medium of SUM149 triple-negative Inflammatory Breast Cancer cell line. These cells, designated as SUM149-MA for metabolic adaptability, are resistant to chemotherapeutic drugs
Kawssar Harb et al.
eLife, 11 (2022-03-10)
In the neocortex, functionally distinct areas process specific types of information. Area identity is established by morphogens and transcriptional master regulators, but downstream mechanisms driving area-specific neuronal specification remain unclear. Here, we reveal a role for RNA-binding proteins in defining
Takashi Irie et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(42), e2307972120-e2307972120 (2023-10-09)
Although generating new neurons in the ischemic injured brain would be an ideal approach to replenish the lost neurons for repairing the damage, the adult mammalian brain retains only limited neurogenic capability. Here, we show that direct conversion of microglia/macrophages
Carla G Silva et al.
Cell, 172(5), 1063-1078 (2018-02-24)
Interneurons navigate along multiple tangential paths to settle into appropriate cortical layers. They undergo a saltatory migration paced by intermittent nuclear jumps whose regulation relies on interplay between extracellular cues and genetic-encoded information. It remains unclear how cycles of pause

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service