Skip to Content
Merck
All Photos(1)

Key Documents

ABE212

Sigma-Aldrich

Anti-Set1A Antibody

serum, from rabbit

Synonym(s):

SET domain containing 1A, Lysine N-methyltransferase 2F, histone-lysine N-methyltransferase SETD1A, Set1/Ash2 histone methyltransferase complex subunit SET1, SET domain-containing protein 1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

technique(s)

ChIP: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SETD1A(9739)

General description

Set1A is a histone methyltransferase (HMT) that is part of the SET1 HMT complex. This protein specifically methylates lysine 4 of histone H3 if the adjacent lysine at position 9 is also unmethylated. Studies demonstrate that Set1A and Set1B localize to largely non-overlapping regions of the chromosome. This suggests Set1A and Set1B bind to unique sets of target genes and that each protien makes non-redundant contributions to the epigenetic control of chromatin structure and gene expression.

Immunogen

GST-tagged recombinant protein corresponding to human Set1A.

Application

Chromatin Immunoprecipitation (ChIP):
A representative lot of this antibody was reported to work in ChIP. See Lee, J.H., et al.(2008). Mol Cell Biol. 28(2):609ā€“618.

Immunocytochemistry:
A representative lot of this antibody was reported to work in immunocytochemistry. See . Lee, J.H., et al. (2007). J Biol Chem. 282(18):13419-13428.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins
Use Anti-Set1A Antibody (Rabbit Polyclonal Antibody) validated in WB to detect Set1A also known as SET domain containing 1A, Set1/Ash2 histone methyltransferase complex subunit SET1.

Quality

Evaluated by Western Blot in PC3 cell lysate.

Western Blot Analysis: 1:1,000 dilution of this antibody detected Set1A on 10 Āµg of PC3 cell lysate.

Target description

~186 kDa observed. A cross-reacting band appears at ~45 kDa

Physical form

Rabbit polyclonal serum containing 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20Ā°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20Ā°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
PC3 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product? Ā 

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service