Skip to Content
Merck
All Photos(1)

Key Documents

AB9078

Sigma-Aldrich

Anti-Ryanodine Receptor 1 Antibody

serum, Chemicon®

Synonym(s):

Anti-CCO, Anti-KDS, Anti-MHS, Anti-MHS1, Anti-PPP1R137, Anti-RYDR, Anti-RYR, Anti-RYR-1, Anti-SKRR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... RYR1(6261)

Specificity

Ryanodine Receptor 1.

Immunogen

Synthetic peptide from the variant TM region of human Ryanodine Receptor 1.

Application

Anti-Ryanodine Receptor 1 Antibody detects level of Ryanodine Receptor 1 & has been published & validated for use in IC, IH & WB.
Western blot: 1:1,000 using ECL. The antibody reacts with the 500 kDa Ryanodine Receptor 1 protein. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Preferred gel percentage is 4-12% gradient gel.

Immunocytochemistry: 1:1,000

Immunohistochemistry: 1:1,000 overnight at 2-8°C using a fluorescently labeled secondary antibody. Suggested fixative is 4% paraformaldehyde in 0.1M PBS (one hour). Suggested permeablization method is 0.05% Triton X-100 in dilution buffer. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS.

Optimal working dilutions must be determined by the end user.

Analysis Note

Control
Western blot = adult mouse skeletal muscle (adult mouse cardiac muscle will be negative).

IHC = adult mouse cerebellum Purkinje cells (adult mouse cerebellum granular cells will be negative).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenli Wang et al.
Fundamental research, 3(3), 459-468 (2022-03-09)
Sweet and umami tastes are elicited by sweet and umami receptors on the tongue and palate epithelium, respectively. However, the molecular machinery allowing the taste reaction remains incompletely understood. Through a phosphoproteomic approach, we identified the key proteins that trigger
Diurnal and nutritional adjustments of intracellular Ca2+ release channels and Ca2+ ATPases associated with restricted feeding schedules in the rat liver.
Baez-Ruiz, A; Cazares-Gomez, K; Vazquez-Martinez, O; Aguilar-Roblero, R; Diaz-Mu?oz, M
Journal of circadian rhythms null
Spaceflight regulates ryanodine receptor subtype 1 in portal vein myocytes in the opposite way of hypertension.
Dabertrand, F; Porte, Y; Macrez, N; Morel, JL
Journal of Applied Physiology (1985)
Peter Braubach et al.
The Journal of general physiology, 144(5), 393-413 (2014-10-29)
Huntington's disease (HD) is caused by an expanded CAG trinucleotide repeat within the gene encoding the protein huntingtin. The resulting elongated glutamine (poly-Q) sequence of mutant huntingtin (mhtt) affects both central neurons and skeletal muscle. Recent reports suggest that ryanodine
Yoshinori Mikami et al.
EBioMedicine, 11, 253-261 (2016-08-22)
Status epilepticus (SE) is a life-threatening emergency that can cause neurodegeneration with debilitating neurological disorders. However, the mechanism by which convulsive SE results in neurodegeneration is not fully understood. It has been shown that epileptic seizures produce markedly increased levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service