Skip to Content
Merck
All Photos(1)

Key Documents

AB770

Sigma-Aldrich

Anti-TIMP-1 Antibody

Chemicon®, from rabbit

Synonym(s):

Tissue Inhibitor of Metalloproteinase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bovine, rat, ferret, sheep, horse, rabbit

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TIMP1(7076)

Specificity

Reacts with TIMP1. Does not react with any other human proteins tested so far. The antibody reacts with native and denatured TIMP1 and TIMP1 in and out of enzyme/inhibitor complex.

Immunogen

Full length purified human fibroblast tissue inhibitor of metalloproteinases {Clark, IM (1991) Matrix 11:76-85}.

Application

Anti-TIMP-1 Antibody detects level of TIMP-1 & has been published & validated for use in ELISA, WB, IH.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western Blotting at 5-10 μg/mL: recognizes a band of approximately 28-30kDa in reduced samples.

Immunohistochemistry: frozen and 4%PFA fixed paraffin sections: 5-20μg/mL enzymatic detection.



Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-1133

Physical form

Purified immunoglobulin. Liquid in PBS containing 0.02% azide.

Storage and Stability

Maintain refrigerated at 2-8°C for up to three months from date of receipt. For extended storage, freeze in convenient undiluted aliquots at -20°C for up to twelve months from date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutsuki Kuraoka et al.
The American journal of pathology, 186(5), 1302-1312 (2016-03-11)
Duchenne muscular dystrophy is a lethal X-linked muscle disorder. We have already reported that osteopontin (OPN), an inflammatory cytokine and myogenic factor, is expressed in the early dystrophic phase in canine X-linked muscular dystrophy in Japan, a dystrophic dog model.
Muraly Puttabyatappa et al.
Reproductive sciences (Thousand Oaks, Calif.), 25(6), 882-892 (2017-03-17)
Prenatal testosterone (T)-treated sheep, similar to women with polycystic ovary syndrome (PCOS), manifests reproductive defects that include multifollicular ovarian phenotype. Women with PCOS manifest increased ovarian matrix metalloproteinases (MMPs) activity. We tested the hypothesis that gestational T excess in sheep
Pedro Ayala et al.
Respiratory research, 19(1), 57-57 (2018-04-11)
Gastric contents aspiration in humans has variable consequences depending on the volume of aspirate, ranging from subclinical pneumonitis to respiratory failure with up to 70% mortality. Several experimental approaches have been used to study this condition. In a model of
Pedro Ayala et al.
American journal of physiology. Lung cellular and molecular physiology, 315(3), L390-L403 (2018-05-11)
Recurrent aspiration of gastric contents has been associated with several interstitial lung diseases. Despite this association, the pathogenic role of aspiration in these diseases has been poorly studied and little is known about extracellular matrix (ECM) changes in animal models
Fanny Huyard et al.
PloS one, 9(4), e92287-e92287 (2014-04-20)
Neonatal high-oxygen exposure leads to elevated blood pressure, microvascular rarefaction, vascular dysfunction and arterial (aorta) rigidity in adult rats. Whether structural changes are present in the matrix of aorta wall is unknown. Considering that elastin synthesis peaks in late fetal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service