Skip to Content
Merck
All Photos(1)

Key Documents

AB16982

Sigma-Aldrich

Anti-Fas Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

FasL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

product name

Anti-Fas Ligand Antibody, Chemicon®, from rabbit

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FAS(355)

Specificity

Human FAS Ligand (FASL). The 13-mer human FASL sequence used for antibody production is 92% (12/13 AA) homologous with rat and mouse FASL. Based upon the sequence homology data, anti-human FASL will likely cross react with the mouse/rat FASL.

Immunogen

A 13 AA peptide sequence mapping near the C-terminus of human FAS ligand (Mita et al. 1994). This peptide is predicted to be extracellular. The peptide was synthesized with C-terminal Cysteine, coupled to KLH.

Application

Anti-Fas Ligand Antibody detects level of Fas Ligand & has been published & validated for use in ELISA & WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting: 1:1,000-1:5,000 Rat, mouse and human FASL have 278, 279, and 281 AA residues, respectively (calculated MW 32kDa). Western blot of the oocyte has detected a specific 31kDa band (Hakuno et al. 1996). A 40 kDa FASL has been detected in mouse T cells (Hahne 1995). An alternate detecting reagent, mFAS-Fc (Suda & Nagata 1994) also detected a cell surface protein of about 40 kDa.

ELISA: 0.5-1μg/mL

Optimal working dilutions must be determined by the end user.

Physical form

Peptide affinity purified immunoglobulin. Liquid in PBS, containing 0.1% BSA.

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prenatal exposure of mice to diethylstilbestrol disrupts T-cell differentiation by regulating Fas/Fas ligand expression through estrogen receptor element and nuclear factor-?B motifs.
Singh, NP; Singh, UP; Nagarkatti, PS; Nagarkatti, M
Journal of Pharmacology and Experimental Therapeutics null
Sisi Wu et al.
Cell biology and toxicology (2021-10-30)
Sirt6, a class III NAD+-dependent deacetylase of the sirtuin family, is a highly specific H3 deacetylase and plays important roles in regulating cellular growth and death. The induction of oxidative stress and death is the critical mechanism involved in cardiomyocyte
Lihong Chen et al.
Immunology and cell biology, 97(8), 714-725 (2019-04-13)
Acute rejection is the major determinant for the long-term survival of donor liver after liver transplantation (LT). The aim of this study was to examine the therapeutic potential of interleukin (IL)-10-FasL-overexpressing immature dendritic cells (imDCs) to induce local immunosuppression in
Elena Beltramo et al.
Acta ophthalmologica, 96(1), e19-e26 (2017-01-28)
Loss of pericytes is one the key events in the pathogenesis of diabetic retinopathy. We have previously demonstrated that human retinal pericytes (HRP) are more vulnerable to intermittent than stable high glucose concentrations, with an increase in apoptosis. Our aim
Yi Peng et al.
Cell death & disease, 11(5), 317-317 (2020-05-07)
Mesenchymal stem cell (MSC) therapy is a promising approach against myocardial infarction (MI). Studies have demonstrated that MSCs can communicate with other cells by secreting exosomes. In the present study, we aimed to identify exosomal microRNAs that might contribute to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service