Skip to Content
Merck
All Photos(1)

Key Documents

AB16502

Sigma-Aldrich

Anti-AIF Antibody, NT

Chemicon®, from rabbit

Synonym(s):

Apoptosis Inducing Factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AIFM1(9131)

Specificity

Apoptosis is characterized by several morphological nuclear changes including chromatin condensation and nuclear fragmentation. These changes are triggered by the activation of members of the caspase family, caspase activated DNase, and several novel proteins (Zamzami & Kroemer 1999). A novel gene, the product of which causes chromatin condensation and DNA fragmentation, was recently identified, cloned, and designated apoptosis inducing factor (AIF) (Susin et al. 1999). Like the critical molecules, cytochrome c and caspase 9, in apoptosis, AIF localizes in mitochondria. AIF translocates to the nucleus when apoptosis is induced and induces mitochondria to release the apoptogenic proteins, cytochrome c and caspase-9. AIF induces chromatin condensation and DNA fragmentation, which are the hallmarks of apoptosis, of the isolated nucleus and the nucleus in live cells by microinjection. AIF is highly conserved between human and mouse and is widely expressed (Susin et al. 1999).

Immunogen

Epitope: N-terminus
Rabbit anti-AIF (N-terminus of mature AIF) polyclonal antibody was raised against a peptide corresponding to amino acids 109 to 122 of human AIF (Susin et al. 1999).

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-AIF Antibody, N-terminus is validated for use in WB for the detection of AIF.
Western blot: 0.25 to 1 μg/mL.

K562 cell lysate from HeLa cells can be used as a positive control and a 67 kDa band should be detected.

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-430

Physical form

Format: Purified
Purified IgG by ion exchange chromatography. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke Zhang et al.
Molecular medicine reports, 12(3), 4415-4421 (2015-06-18)
Sepsis is one of the leading causes of mortality in severe systemic inflammatory syndrome. The endotoxin-induced inflammatory response has been linked to the development of sepsis. Rhein is a lipophilic anthraquinone isolated from Rheum rhabarbarum (rhubarb), which has a protective
Keyu Ren et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 51(6), e7065-e7065 (2018-04-26)
Ulcerative colitis is a chronic inflammatory disease of the colon where intestinal motility is disturbed. Interstitial cells of Cajal (ICC) are required to maintain normal intestinal motility. In the present study, we assessed the effect of tumor necrosis factor-alpha (TNF-α)
Zhonghua Cheng et al.
Bioengineered, 13(1), 1359-1376 (2022-01-12)
Gastric cancer (GC), one of the most prevalent malignancies across the world, has an increasing incidence rate. Long non-coding RNA (lncRNA) PURPL (also referred to as LINC01021) has been demonstrated to influence malignant GC behaviors and partake in other cancers.
Qinqin Lin et al.
Medical science monitor basic research, 22, 165-174 (2016-12-17)
BACKGROUND Aquaporin-2 (AQP2) plays a major role in water reabsorption in the renal collecting duct, and is involved in a variety of renal disease. Recent studies have indicate that sirtuin1 (SIRT1) exerts renoprotective properties against kidney diseases. This study aimed
Yong Wang et al.
Molecular medicine (Cambridge, Mass.), 25(1), 17-17 (2019-05-08)
Obesity is one of the leading causes of insulin resistance. Accumulating reports have highlighted that serum amyloid A-1 (SAA1) is a potential candidate that is capable of attenuating insulin resistance. Hence, we conducted the current study with aims of investigating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service