Skip to Content
Merck
All Photos(3)

Key Documents

AB1585

Sigma-Aldrich

Anti-Dopamine β Hydroxylase Antibody

serum, Chemicon®

Synonym(s):

DBH, Dopamine beta-monooxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

bovine, guinea pig, rat, human, feline, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
radioimmunoassay: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DBH(1621)

Specificity

Recognizes Dopamine beta hydroxylase. AB1585 reacts with a single band on Western blots of bovine adrenal homogenates and stains only cells known to contain DBH, such as sympathetic neurons, adrenal medullary cells and central adrenergic neurons. Staining is abolished by preincubation with enzyme.

Immunogen

Native enzyme from bovine adrenal glands.

Application

Detect Dopamine β Hydroxylase using this Anti-Dopamine β Hydroxylase Antibody validated for use in IH, FUNC, RIA & WB.
Immunohistochemistry: 1:2,000 on rabbit and guinea pig using an ABC detection method.

Western blot

RIA

Inhibitory studies in vivo and in vitro.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Target description

69 kDa

Linkage

Replaces: AB145

Physical form

Rabbit Serum. Lyophilized, no preservatives. Reconstitute with 50 μL of sterile distilled water. Centrifuge to remove any insoluble material.
Unpurified

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20 to -70°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles. Glycerol can be added 1:1 for additional stability.

Analysis Note

Control
Brain (Striatum/Substantia Nigra)

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Buffalari et al.
Neuroscience, 281, 241-250 (2014-07-23)
Central noradrenergic (NA) signaling contributes critically to multiple behavioral effects of cocaine administration, particularly stress- and anxiety-related effects. The present study examined the ability of acute cocaine to induce the immediate early gene product, cFos, in NA neurons and stress-related
Junchao Tong et al.
Journal of neurochemistry, 102(5), 1691-1702 (2007-05-09)
We recently identified a noradrenaline-rich caudomedial subdivision of the human nucleus accumbens (NACS), implying a special function for noradrenaline in this basal forebrain area involved in motivation and reward. To establish whether the NACS, as would be expected, contains similarly
Directed differentiation of neural cells to hypothalamic dopaminergic neurons.
Ohyama, K; Ellis, P; Kimura, S; Placzek, M
Development null
Jian Qi et al.
PloS one, 6(8), e23275-e23275 (2011-08-23)
The synaptic connections between neurokinin 1 (NK1) receptor-like immunoreactive (LI) neurons and γ-aminobutyric acid (GABA)-, glycine (Gly)-, serotonin (5-HT)- or dopamine-β-hydroxylase (DBH, a specific marker for norepinephrinergic neuronal structures)-LI axon terminals in the rat medullary dorsal horn (MDH) were examined
Jimena Baleriola et al.
Scientific reports, 6, 25928-25928 (2016-05-14)
Programmed cell death occurs naturally at different stages of neural development, including neurogenesis. The functional role of this early phase of neural cell death, which affects recently differentiated neurons among other cell types, remains undefined. Some mouse models defective in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service