Skip to Content
Merck
All Photos(1)

Key Documents

AB1566

Sigma-Aldrich

Anti-Substance P Antibody, pain

serum, Chemicon®

Synonym(s):

Anti-Hs.2563, Anti-NK2, Anti-NKNA, Anti-NPK, Anti-TAC2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
radioimmunoassay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TAC1(6863)

General description

Substance P is a tachykinin family neuropeptide that functions as a neurotransmitter and neuromodulator. In the CNS, Substance P is involved in the regulation of mood, anxiety, sleep, saiety, reinforcement, respiratory rhythm and pain processing. In nociception, the neuropeptide is released at local spinal cord synapse. Recent research now shows substance P is potentially involved in endothelial cell differentiation as well as diverse roles in neuroinflammatory response. The endogenous receptor for Substance P is neurokinin 1 receptor (NK1 receptor), which belongs to the tachykinin receptor sub-family of GPCRs.

Specificity

Substance P

Application

RIA:
1:10,000 dilution from a previous lot was used in radioimmunioassay. 80% binding detects at least 5 pg/mL.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuroinflammation & Pain
This Anti-Substance P Antibody, pain is validated for use in IH(P), RIA for the detection of Substance P.

Quality

Immunohistochemistry(paraffin):
Substance P (AB1566) representative staining pattern/morphology in Alzheimer’s brain. Tissue pretreated with Citrate pH6.0, antigen retrieval. Antibody diluted to 1:100.
IHC-Paraffin Staining With Citrate pH 6.0 Epitope Retrieval: Alzheimer’s Brain.

Physical form

Rabbit polyclonal serum containing 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Spinal Cord.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sally I Firth et al.
Visual neuroscience, 19(4), 531-540 (2003-01-04)
High levels of endogenous cholecystokinin (CCK) are present in the rat retina (Eskay & Beinfeld, 1982), but the cellular localization and physiological actions of CCK in the rat retina are uncertain. The goals of this study were to characterize the
Zhijing Wang et al.
Journal of inflammation (London, England), 14, 13-13 (2017-06-13)
Traumatic brain injury (TBI) is a public health issue. As the major complaint in 51% of TBI patients, chronic pain is an important aspect in TBI treatment. Tetramethylpyrazine (TMP) is an important compound in Ligustrazine, an analgesic drug in traditional
Kenzen Kou et al.
Acta dermato-venereologica, 92(5), 521-528 (2012-05-09)
Pruritus is a common symptom of psoriasis, which affects quality of life. This symptom accompanies the hyper-innervation of sensory C-fibres in psoriatic lesions. Two extracellular molecules, nerve growth factor (NGF) and semaphorin-3A, regulate C-fibre extension. In this study, the expression
Aristea Sideri et al.
Cellular and molecular gastroenterology and hepatology, 1(4), 420-432 (2015-11-07)
Substance P (SP), neurokinin-1 receptors (NK-1Rs) are expressed in mesenteric preadipocytes and SP binding activates proinflammatory signalling in these cells. We evaluated the expression levels of SP (Tac-1), NK-1R (Tacr-1), and NK-2R (Tacr-2) mRNA in preadipocytes isolated from patients with
Sai Y Veruva et al.
Clinical orthopaedics and related research, 475(5), 1369-1381 (2016-08-05)
The pathophysiology and mechanisms driving the generation of unintended pain after total disc replacement (TDR) remain unexplored. Ultrahigh-molecular-weight polyethylene (UHMWPE) wear debris from TDRs is known to induce inflammation, which may result in pain. The purpose of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service