Skip to Content
Merck
All Photos(3)

Key Documents

AB15299-I

Sigma-Aldrich

Anti-Dendrin

from rabbit, purified by affinity chromatography

Synonym(s):

DDN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
human, mouse, rat
technique(s):
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DDN(23109)

General description

Dendrin (UniProt: O94850) is encoded by the DDN (also known as KIAA0749) gene (Gene ID: 23109) in human. Dendrin is a proline-rich cytoplasmic protein that was originally recognized in telencephalic dendrites of sleep-deprived rats. Dendrin is specifically expressed in the brain and in kidneys. In the brain, it interacts with alpha-actinin and possibly modulates the structure of the actin cytoskeleton. In kidney it plays a role in apoptosis of kidney glomerular podocytes. Dendrin contains two putative nuclear localization signals (NLSs) and three PPXY motifs that are conserved among human, rat, and mouse. Dendrin is reported to interact with alpha-actinin and Maguk with inverted orientation (MAGI) or synaptic scaffolding molecule (S-SCAM). Interaction with S-SCAM is shown to be responsible for the retention of dendrin in the cytoplasm of mammalian cells and prevent its nuclear import. In experimental glomerulonephritis, dendrin is shown to relocate from the slit diaphragms (SD) to the nucleus of injured podocytes. High-doses of TGF- 1 can directly promote the import of dendrin into the nucleus and in the nucleus Dendrin enhances staurosporine- and TGF- 1-mediated apoptosis. Acute nicotine treatment is reported to induce a rapid, but short-lived, expression of the dendritically targeted, corticolimbic mRNA dendrin in specific regions of adolescent brain. (Ref.: Schochet, TL et al (2008). Eur.J. Neurosci. 28(10):1967-79; Kremerskothen, J et al. (2006). J. Neurochem. 96(6):1659-66; Asanuma, K et al. (2007). Proc. Natl. Acad. Sci. USA. 104(24): 10134-39).

Specificity

This polyclonal antibody recognizes Dendrin in human, mouse, and rat. It targets an epitope of 17 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of human Dendrin.

Application

Anti-Dendrin Antibody, Cat. No. AB15299-I, is a highly specific rabbit polyclonal antibody that targets Dendrin and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: 2 µg/mL from a representative lot detected Dendrin in 10 µg of human hippocampus tissue lysate.

Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Dendrin in rat cerebral cortex, mouse cerebellum, and rat kidney tissue.

Quality

Evaluated by Western Blotting in mouse hippocampus tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Dendrin in 10 µg of mouse hippocampus tissue lysate.

Target description

~70 kDa observed, 76 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: AB15299

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marija Jurić et al.
Neuropeptides, 86, 102126-102126 (2021-02-02)
Chronic stress has various effects on organisms and is sex-specific. The aim of the study was to describe the expression of synapse strengthening protein, dendrin, in the spinal cord (SC) and the dependence of its expression on chronic stress and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service