967106
Stainless steel clamp for filling nozzle attachment to stainless steel drums
Synonym(s):
Clamp for filling nozzle, drums
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122409
NACRES:
NA.03
Recommended Products
shipped in
ambient
storage temp.
room temp
General description
Safety and convenient handling are the basis for an important part of our very special product developments. When using a pressurizing withdrawal for stainless steel drums our filling nozzle (9.67100.1090 / 9.67100.9090) can be stored safely and directly on the drum by using a screw clamp.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productās label following the words āLotā or āBatchā.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service