Skip to Content
Merck
All Photos(1)

Key Documents

8.40110

Sigma-Aldrich

Isooctylthioglycolate

for synthesis

Synonym(s):

Isooctylthioglycolate, Mercaptoacetic acid isooctyl ester, Isooctyl mercaptoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O2S
CAS Number:
Molecular Weight:
204.33
MDL number:
UNSPSC Code:
12352108
EC Index Number:
246-613-9
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.3 hPa ( 20 °C)

Quality Level

form

liquid

potency

348 mg/kg LD50, oral (Rat)

mp

<-50 °C

transition temp

flash point >94.4 °C

density

0.97 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCC(=O)OCCCCCC(C)C

InChI

1S/C10H20O2S/c1-9(2)6-4-3-5-7-12-10(11)8-13/h9,13H,3-8H2,1-2H3

InChI key

RZBBHEJLECUBJT-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 0.970 - 0.975
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

201.9 °F - closed cup

Flash Point(C)

> 94.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service