Skip to Content
Merck
All Photos(2)

Key Documents

8.20350

Sigma-Aldrich

Tributyl citrate

greener alternative

for synthesis

Synonym(s):

Tributyl citrate, Citric acid tributyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOCH2CH2CH2CH3)(CH2COOCH2CH2CH2CH3)2
CAS Number:
Molecular Weight:
360.44
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-071-2
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

liquid

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

169-171 °C/1 hPa

mp

-20 °C

transition temp

flash point 184 °C

density

1.04 g/cm3 at 20 °C

greener alternative category

storage temp.

2-30°C

InChI

1S/C18H32O7/c1-4-7-10-23-15(19)13-18(22,17(21)25-12-9-6-3)14-16(20)24-11-8-5-2/h22H,4-14H2,1-3H3

InChI key

ZFOZVQLOBQUTQQ-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product can replace highly toxic diluents in membrane preparation and thus aligns with "Less Hazardous Chemical Syntheses". Click here for more information.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.042 - 1.045
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service