Skip to Content
Merck
All Photos(1)

Key Documents

8.18289

Sigma-Aldrich

(+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxy-α-D-erythro-hex-2- enopyranoside

for synthesis

Synonym(s):

(+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxy-α-D-erythro-hex-2- enopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
258.27
UNSPSC Code:
12352108
NACRES:
NA.22

Quality Level

form

powder

mp

76-78 °C

bulk density

200 kg/m3

storage temp.

2-30°C

InChI

1S/C12H18O6/c1-4-15-12-6-5-10(17-9(3)14)11(18-12)7-16-8(2)13/h5-6,10-12H,4,7H2,1-3H3

InChI key

OSPOBRCWZOKNRC-UHFFFAOYSA-N

General description

for synthesis

Application

(+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxy-α-D-erythro-hex-2- enopyranoside for synthesis. CAS 3323-72-6, molar mass 258.27 g/mol.

Analysis Note

Assay (DSC)≥ 99.0 %Melting range (lower value)≥ 77 °CMelting range (upper value)≤ 80 °CSpec. rotation [α²0/D (c=1 in toluene)+105.0 - +107.0 °Identity (IR)passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service