Skip to Content
Merck
All Photos(1)

Key Documents

8.17500

Sigma-Aldrich

Sodium butyrate

special quality

Synonym(s):

Sodium butyrate, Butyric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NaO2
CAS Number:
Molecular Weight:
110.09
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-857-6
NACRES:
NA.22
grade:
for analytical purposes
form:
powder
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

form

powder

mp

244 °C

density

1.324 g/cm3 at 30 °C

storage temp.

2-30°C

SMILES string

[Na+].[O-]C(=O)CCC

InChI

1S/C4H8O2.Na/c1-2-3-4(5)6;/h2-3H2,1H3,(H,5,6);/q;+1/p-1

InChI key

MFBOGIVSZKQAPD-UHFFFAOYSA-M

Analysis Note

Assay (HClO₄): ≥ 98.5 %
Identity (IR): passes test
Appearance (white powder): passes test
Free acid/base: ≤ 2.0 ml
Heavy metals (Pb): ≤ 0.0005 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service