Skip to Content
Merck
All Photos(1)

Key Documents

8.14312

Sigma-Aldrich

Acenaphthene

for synthesis

Synonym(s):

Acenaphthene, 1,8-Ethylenenaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10
CAS Number:
Molecular Weight:
154.21
MDL number:
UNSPSC Code:
12352002
EC Index Number:
201-469-6
NACRES:
NA.22
Assay:
≥96.0% (GC)

vapor pressure

<0.005 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

autoignition temp.

>450 °C

potency

>16000 mg/kg LD50, oral (Rat)

bp

278 °C/1013 hPa

mp

95 °C

transition temp

flash point 135 °C

density

1.15 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1Cc2c3c(ccc2)cccc31

InChI

1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2

InChI key

CWRYPZZKDGJXCA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 91 °C
Melting range (upper value): ≤ 95 °C
Identity (IR): passes test

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service