Skip to Content
Merck
All Photos(1)

Key Documents

8.06806

Sigma-Aldrich

2-Nitrotoluene

for synthesis

Synonym(s):

2-Nitrotoluene, o-Nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(NO2)C6H4CH3
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352102
EC Index Number:
201-853-3
NACRES:
NA.22
form:
oil
Assay:
≥99.0% (GC)

vapor pressure

0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

oil

autoignition temp.

420 °C

potency

891 mg/kg LD50, oral (Rat)

expl. lim.

1.47-8.8 % (v/v)

pH

6-8 ( in H2O)

bp

221-222 °C/1013 hPa

mp

-9 °C

transition temp

flash point 95 °C

solubility

0.65 g/L (experimental)

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[N+](=O)([O-])c1c(cccc1)C

InChI

1S/C7H7NO2/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3

InChI key

PLAZTCDQAHEYBI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.162 - 1.163
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Muta. 1B - Repr. 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

203.0 °F

Flash Point(C)

95 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service