Skip to Content
Merck
All Photos(2)

Key Documents

8.00763

Sigma-Aldrich

D-(+)-Malic acid

for the resolution of racemates for synthesis

Synonym(s):

D-(+)-Malic acid, (R)-(+)-Malic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH(OH)CH2COOH
CAS Number:
Molecular Weight:
134.09
MDL number:
UNSPSC Code:
12352106
EC Index Number:
211-262-2
NACRES:
NA.22

Quality Level

Assay

≥99.0% (acidimetric)

pH

2.2 (20 °C, 10 g/L in H2O)

mp

101-104 °C

solubility

364 g/L

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)C(O)CC(=O)[O-].[H+].[H+]

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)

InChI key

BJEPYKJPYRNKOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 101 °C
Melting range (upper value): ≤ 104 °C
Spec. rotation [α²0/D (c=5 in pyridine): +27.0 - +30.0 °
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service