Skip to Content
Merck
All Photos(1)

Key Documents

8.00640

Sigma-Aldrich

2-Hydroxybenzaldehyde

for synthesis

Synonym(s):

2-Hydroxybenzaldehyde, Salicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O2
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352114
EC Index Number:
201-961-0
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.79 hPa ( 25 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

potency

520 mg/kg LD50, oral (Rat)
3000 mg/kg LD50, skin (Rabbit)

pH

6-8 (20 °C in H2O)

mp

-7 °C

transition temp

flash point 78 °C

density

1.17 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1c(cccc1)C=O

InChI

1S/C7H6O2/c8-5-6-3-1-2-4-7(6)9/h1-5,9H

InChI key

SMQUZDBALVYZAC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99 %
Density (d 20 °C/ 4 °C): 1.165 - 1.167
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service