676777
(±)-Verapamil, Hydrochloride
A papaverine derivative that blocks Ca2+ channels (principally the L-type) in smooth and cardiac muscle cells.
Synonym(s):
(±)-Verapamil, Hydrochloride, L-Type Calcium Channel Blocker V
About This Item
Recommended Products
Quality Level
description
Merck USA index - 14, 9950
Assay
≥99% (perchloric acid titration)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
methanol: 50 mg/mL
water: 70 mg/mL
ethanol: soluble
shipped in
ambient
storage temp.
10-30°C
SMILES string
[Cl-].[N+H](CCCC(C(C)C)(c2cc(c(cc2)OC)OC)C#N)(CCc1cc(c(cc1)OC)OC)C
InChI
1S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20H,8,13-16H2,1-7H3;1H
InChI key
DOQPXTMNIUCOSY-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Warning
Other Notes
Shchepotin, I.B., et al. 1994. Anticancer Res.14, 1027.
Janis, R., et al. 1987. Adv. Drug Res.16, 309.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Chronic 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service