Skip to Content
Merck
All Photos(1)

Key Documents

580218

Millipore

Taurocholic Acid, Sodium Salt

ULTROL® Grade, ≥98% (TLC), detergent to solubilize membrane-bound proteins in their native state

Synonym(s):

Taurocholic Acid, Sodium Salt, ULTROL® Grade, Sodium Taurocholate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44NO7S · Na
CAS Number:
Molecular Weight:
537.68
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.21
Pricing and availability is not currently available.

description

Merck USA index - 14, 9075

Quality Level

Assay

≥98% (TLC)

form

solid

mol wt

micellar wt 2100

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

aggregation number

4

color

white

pKa 

1.9

CMC

3 - 11 mM

solubility

water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Na+].[S](=O)(=O)([O-])CCNC(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@H](C[C@H]3O)C[C@@H](CC4)O)C)C[C@@H]2O)CC1)C)C

InChI

1S/C26H45NO7S.Na/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29;/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34);/q;+1/p-1/t15-,16+,17-,18-,19+,20+,21-,22+,24+,25+,26-;/m1./s1

InChI key

JAJWGJBVLPIOOH-IZYKLYLVSA-M

General description

Absorbance (100 mM, H2O, 280 nm): <0.30.
Highly purified and intended for use as a detergent to solubilize membrane-bound proteins in their native state (absorbance: <0.30 at 280 nm, 100 mM, H2O; aggregation number: 4; CMC: 3-11 mM; micellar weight: 2100; pKa: 1.9).
Highly purified and intended for use as a detergent to solubilize membrane-bound proteins in their native state. Aggregation number: 4.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and refrigerate (4°C ). Stock solutions are stable for up to 2 months at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service