Skip to Content
Merck
All Photos(1)

Key Documents

5.43841

Supelco

Potassium phosphate monobasic

for HPLC LiChropur

Synonym(s):

Potassium phosphate monobasic, mono-Potassium orthophosphate, Potassium biphosphate, Potassium phosphate monobasic, Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
MDL number:
UNSPSC Code:
41116105
EC Index Number:
231-913-4
NACRES:
NA.21
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Assay

≥99.5% (acidimetric)

form

solid

technique(s)

HPLC: suitable

pH

4.2-4.6 (20 °C, 20 g/L in H2O)

mp

252.6 °C (lit.)

solubility

208 g/L

density

2.338 g/mL at 25 °C (lit.)

bulk density

1200 kg/m3

storage temp.

2-30°C

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

  • Unlocking the potential of biochar: Potassium phosphate monobasic is utilized to enhance the adsorption capacity of biochar composites for removing heavy metals like Pb(II) and Cd(II) from water, highlighting its effectiveness in environmental cleanup strategies (Li et al., 2024).
  • Partitioning recombinant chitinase from Nicotiana benthamiana by an aqueous two-phase system: Demonstrates the application of potassium phosphate monobasic in aqueous two-phase systems for efficient partitioning of enzymes, which is crucial for biotechnological and industrial applications (Tue et al., 2024).
  • Unlocking Nature′s Toolbox: This study showcases how potassium phosphate monobasic can be used to optimize conditions for the production of recombinant proteins, enhancing yields and streamlining production processes in biomanufacturing (Rajak et al., 2024).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Separation and identification of oligonucleotides differing by one base in length using Size Exclusion Chromatography combined with multi-angle light scattering (SEC-MALS) detector.

A new RP-HPLC method is proposed for the determination of organic acids in 100% aqueous mobile phase using an Ascentis® Express AQ-C18 column

Protocols

Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.

Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.

Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.

Powdered black pepper tested for pierine content follows USP guidelines using HPTLC and HPLC for dietary supplements.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service