Skip to Content
Merck
All Photos(1)

Key Documents

524632

Millipore

Phosphatase Inhibitor Cocktail V

50X, lyophilized solid, for the inhibition of acid, alkaline, serine/threonine and protein tyrosine phosphatases. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Phosphatase inhibitor cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54
form:
lyophilized solid
solubility:
water: soluble
storage temp.:
2-8°C
Pricing and availability is not currently available.

Product Name

Phosphatase Inhibitor Cocktail Set V, 50X, Lyophilized, The Phosphatase Inhibitor Cocktail Set V, 50X, Lyophilized controls the activity of Phosphatase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

A cocktail of four phosphatase inhibitors for broad-spectrum inhibition of acid, alkaline, serine/threonine and protein tyrosine phosphatases. Available as a 1 ml vial or a set of five 1 ml vials. Each vial, when reconstituted with 1 ml H2O contains 250 mM Sodium Fluoride, 50 mM β-Glycerophosphate (Cat. No. 35675), 50 mM Sodium Pyrophosphate Decahydrate, 50 mM Sodium Orthovanadate.

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C) for long-term storage. Dilute 1:50 just prior to use.
Reconstitute each vial with 1 ml H₂O.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bernd Stratmann et al.
International journal of molecular sciences, 23(13) (2022-07-10)
An oversupply of nutrients with a loss of metabolic flexibility and subsequent cardiac dysfunction are hallmarks of diabetic cardiomyopathy. Even if excess substrate is offered, the heart suffers energy depletion as metabolic fluxes are diminished. To study the effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service