Skip to Content
Merck
All Photos(1)

Key Documents

5.06188

Sigma-Aldrich

Sertraline Hydrochloride

Synonym(s):

Sertraline Hydrochloride, Serotonin (5-HT) Reuptake Inhibitor, Sertraline, Zoloft

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17Cl2N · xHCl
CAS Number:
Molecular Weight:
306.23 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

potency

3.3 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mM
ethanol: 50 mM

storage temp.

2-8°C

SMILES string

[Cl-].Clc1c(ccc(c1)[C@@H]2CC[C@@H](c3c2cccc3)[N+H2]C)Cl

InChI

1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1

InChI key

BLFQGGGGFNSJKA-XHXSRVRCSA-N

General description

A very potent uptake inhibitor highly selective for serotonin (Ki = 3.3 nM). Clinically prescribed as an antidepressant drug. Often used in studying major depressive disorders.

Biochem/physiol Actions

Primary Target
5-HT uptake transporter

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Brunoni, R., et al. 2013. JAMA Psychiatry.70, 383.

Kobayashi, K., et al. 1995. Br. J. Clin. Pharmacol.40, 481.
Heym, J., et al. 1988. J. Clin. Psychiatry.49 Suppl, 40.
Koe, K., et al. 1983. J. Pharmacol. Exp. Ther.226, 700.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service