Skip to Content
Merck
All Photos(1)

Key Documents

5.05024

Sigma-Aldrich

Ibotenic Acid

Synonym(s):

Ibotenic Acid, 2-amino-2-(3-hydroxyisoxazol-5-yl) acetic acid, α-Amino-(3-hydroxy-5-isoxazolyl)acet­ic acid, Glutamate Receptor Agonist, Ibotenic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 10 mM

storage temp.

−20°C

SMILES string

[N+H3]C(c1[o]nc(c1)O)C(=O)[O-]

InChI

1S/C5H6N2O4/c6-4(5(9)10)2-1-3(8)7-11-2/h1,4H,6H2,(H,7,8)(H,9,10)

InChI key

IRJCBFDCFXCWGO-UHFFFAOYSA-N

General description

A highly potent agonist of NMDA and mGlu receptors. A frequently used brain-lesioning agent inducing excitotoxic lesions of neurons without damaging crossing fiber tracts.

Warning

Toxicity: Regulatory Review (Z)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Izquierdo, A., et al. 2013. J. Neurosci.33, 4105.
Rhodes, V., et al. 2013. J. Neurosci.33, 3380.
Yu, X., et al. 2012. Pharmacol. Biochem. & Beh.101, 479.
Albasser, M., et al. 2012. Beh. Neurosci.126, 659.
Eijkenboom, M., et al. 2000. Neurosci.101, 27.
Isacson, O., et al. 1984. Nature.311, 458.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service